Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 4 | 927 | ||
NM_019804.4 | NP_062778.2 | 4 | 860 | |||
GenBank mRNA | AF142672.1 | - | 4 | 837 | ||
AF158746.1 | - | 4 | 839 | |||
AK018658.1 | - | 4 | 824 | |||
AK041692.1 | - | 4 | 852 | |||
AK045602.1 | - | 4 | 832 | |||
AK050005.1 | - | 4 | 856 | |||
AK052013.1 | - | 4 | 854 | |||
AK053603.1 | - | 4 | 921 | |||
AK085368.1 | - | 4 | 846 | |||
AK136637.1 | - | 4 | 772 | |||
BC013492.1 | - | 4 | 823 | 4218187 | ||
BC031115.1 | - | 4 | 912 | 5026168 |