Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 2 | 367 | ||
NM_019804.4 | NP_062778.2 | 2 | 300 | |||
GenBank mRNA | AF142672.1 | - | 2 | 277 | ||
AF158746.1 | - | 2 | 279 | |||
AK018658.1 | - | 2 | 264 | |||
AK041692.1 | - | 2 | 292 | |||
AK045602.1 | - | 2 | 272 | |||
AK050005.1 | - | 2 | 296 | |||
AK052013.1 | - | 2 | 294 | |||
AK053603.1 | - | 2 | 361 | |||
AK085368.1 | - | 2 | 286 | |||
AK136637.1 | - | 2 | 212 | |||
BC013492.1 | - | 2 | 263 | 4218187 | ||
BC031115.1 | - | 2 | 352 | 5026168 |