Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019824.3 | NP_062798.1 | 5, 6 | 436 | ||
GenBank mRNA | AB038243.1 | - | 4, 5 | 372 | ||
AK003485.1 | - | 5, 6 | 467 | |||
AK005437.1 | - | 5, 6 | 455 | |||
AK171651.1 | - | 5, 6 | 451 | |||
BC013618.1 | - | 5, 6 | 436 | 4013537 | ||
BC054440.1 | - | 5, 6 | 403 | 6395280 | ||
BC055036.1 | - | 5, 6 | 403 |