Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167799.1 | NP_001161271.1 | 9, 10 | 959 | ||
NM_019829.4 | NP_062803.4 | 8, 9 | 1258 | |||
GenBank mRNA | AF232709.1 | - | 8, 9 | 626 | ||
AK166087.1 | - | 8, 9 | 1261 | |||
BC004849.1 | - | 9, 10 | 892 | 2655845 | ||
BC021883.1 | - | 8, 9 | 1241 | 3487476 | ||
DV649852.1 | - | 3, 4 | 222 |