Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168505.1 | NP_001161977.1 | 2 | 849 | ||
NM_019658.6 | NP_062632.2 | 2 | 890 | |||
GenBank mRNA | AF068921.1 | - | 1 | 525 | ||
AK146447.1 | - | 2 | 887 | |||
BC026364.1 | - | 1 | 756 | 4014650 | ||
BC049775.1 | - | 1 | 755 | 6515905 | ||
BC083060.1 | - | 2 | 849 | 5356514 |