Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028920.1 | - | 14 | 2020 | ||
AK004801.1 | - | 14 | 2026 | |||
AK089092.1 | - | 14 | 2033 | |||
BC042602.1 | - | 14 | 2903 | |||
BC054433.1 | - | 12 | 2600 | |||
BC060170.1 | - | 14 | 2900 | |||
BC060209.1 | - | 13 | 2516 | |||
BC078448.1 | - | 13 | 2406 | |||
BC125521.1 | - | 14 | 2016 | 40130036 | ||
BC137746.1 | - | 14 | 2016 | 8860765 |