Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019825.3 | NP_062799.2 | 10 | 4009 | ||
GenBank mRNA | AF216186.1 | - | 9 | 3894 | ||
AK129079.1 | - | 3 | 1168 | |||
AK142566.1 | - | 4 | 2194 | |||
AK171864.1 | - | 10 | 4167 | |||
BC083187.1 | - | 10 | 4009 | 30544739 | ||
JF707635.1 | - | 9 | 3868 |