Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc4ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163452.1 | NP_001156924.1 | 15 | 1816 | ||
NM_019828.2 | NP_062802.2 | 15 | 1792 | |||
GenBank mRNA | AF093589.1 | - | 11 | 1288 | ||
AF130458.1 | - | 15 | 1740 | |||
AF509881.1 | - | 15 | 1738 | |||
AK156047.1 | - | 15 | 1765 | |||
AK170085.1 | - | 15 | 1754 | |||
AK171404.1 | - | 15 | 1765 | |||
AK186727.1 | - | 2 | 130 | |||
AK188595.1 | - | 2 | 130 | |||
AK190938.1 | - | 2 | 130 | |||
AK200696.1 | - | 2 | 131 | |||
AK220239.1 | - | 15 | 1792 | |||
BC003931.1 | - | 8 | 752 | |||
BC033274.1 | - | 15 | 1744 | 3673725 | ||
BC057330.1 | - | 15 | 1774 | 6816155 |