Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291071.1 | NP_001278000.1 | 2 | 260 | ||
NM_019703.4 | NP_062677.1 | 2 | 260 | |||
GenBank mRNA | AF123533.1 | - | 2 | 225 | ||
AK004790.1 | - | 2 | 215 | |||
AK020367.1 | - | 2 | 215 | |||
AK049841.1 | - | 2 | 214 | |||
AK054193.1 | - | 2 | 215 | |||
AK076803.1 | - | 2 | 215 | |||
AK078254.1 | - | 2 | 215 | |||
AK098162.1 | - | 2 | 214 | |||
AK152670.1 | - | 2 | 215 | |||
AK154125.1 | - | 2 | 215 | |||
AK165422.1 | - | 2 | 214 | |||
AK165425.1 | - | 2 | 214 | |||
AK170624.1 | - | 2 | 215 | |||
AK171062.1 | - | 2 | 215 | |||
AK184036.1 | - | 1 | 42 | |||
AY779275.1 | - | 2 | 177 | |||
AY779276.1 | - | 2 | 177 | |||
BC006926.1 | - | 2 | 199 | 2655383 | ||
BY243493.1 | - | 2 | 260 | |||
CT010268.1 | - | 2 | 168 | |||
Y19008.1 | - | 2 | 217 |