Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291229.1 | NP_001278158.1 | 9 | 1674 | ||
NM_019765.5 | NP_062739.2 | 9 | 1674 | |||
GenBank mRNA | AK014540.1 | - | 9 | 1445 | ||
AK029945.1 | - | 9 | 1511 | |||
AK220172.1 | - | 9 | 1635 | |||
BC007191.1 | - | 9 | 1614 | 3592614 | ||
BC029243.1 | - | 6 | 796 | |||
BC052632.1 | - | 7 | 1080 |