Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019767.2 | NP_062741.1 | 4, 5 | 532 | ||
GenBank mRNA | AB024984.1 | - | 4, 5 | 532 | ||
AK002475.1 | - | 4, 5 | 495 | |||
AK076107.1 | - | 4, 5 | 520 | |||
AK145663.1 | - | 4, 5 | 523 | |||
BC001988.1 | - | 4, 5 | 510 | 3485409 |