Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019785.2 | NP_062759.2 | 5 | 483 | ||
GenBank mRNA | AF190797.1 | - | 5 | 450 | ||
AK161642.1 | - | 5 | 443 |