Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019794.4 | NP_062768.1 | 3 | 281 | ||
GenBank mRNA | AB028853.1 | - | 3 | 282 | ||
AK031858.1 | - | 3 | 348 | |||
AK077672.1 | - | 3 | 352 | |||
AK083208.1 | - | 3 | 349 | |||
AK163255.1 | - | 3 | 280 | |||
AK169169.1 | - | 3 | 341 |