Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019739.3 | NP_062713.2 | 2 | 1800 | ||
GenBank mRNA | AF114258.1 | - | 2 | 1718 | ||
AF126056.1 | - | 2 | 1341 | |||
AJ252157.1 | - | 2 | 1763 | |||
AK053633.1 | - | 1 | 500 | |||
AK137629.1 | - | 2 | 1748 | |||
AK154041.1 | - | 2 | 1801 |