Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285891.1 | NP_001272820.1 | 8, 9 | 1104 | ||
NM_019748.3 | NP_062722.1 | 8, 9 | 1104 | |||
GenBank mRNA | AB024303.1 | - | 8, 9 | 992 | ||
AK010313.1 | - | 8, 9 | 981 | |||
AK011783.1 | - | 5, 6 | 451 | |||
AK087556.1 | - | 8, 9 | 995 | |||
AK090012.1 | - | 8, 9 | 991 | |||
AK154139.1 | - | 8, 9 | 1017 | |||
AK159672.1 | - | 8, 9 | 1006 | |||
BC068164.1 | - | 8, 9 | 993 | 30359189 |