Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019699.1 | NP_062673.1 | 5 | 713 | ||
GenBank mRNA | AF126798.1 | - | 5 | 713 | ||
AK083282.1 | - | 680 | ||||
AK143524.1 | - | 731 | ||||
BC057189.1 | - | 5 | 701 | 5137343 | ||
HQ264057.1 | - | 5 | 606 | |||
HQ264058.1 | - | 5 | 606 |