Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019777.3 | NP_062751.2 | 7, 8 | 1036 | ||
GenBank mRNA | AB016589.1 | - | 5, 6 | 724 | ||
AK088580.1 | - | 7, 8 | 1038 | |||
AK149911.1 | - | 7, 8 | 1025 | |||
AK153046.1 | - | 7, 8 | 1037 | |||
BC037446.1 | - | 6, 7 | 708 | 3980145 | ||
CT010206.1 | - | 5, 6 | 690 | |||
CT010313.1 | - | 5, 6 | 618 |