Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204165.1 | NP_001191094.1 | 23 | 2268 | ||
NM_001204166.1 | NP_001191095.1 | 20 | 2160 | |||
NM_019688.2 | NP_062662.1 | 22 | 2214 | |||
GenBank mRNA | AB021132.1 | - | 23 | 2243 | ||
AB037668.1 | - | 14 | 1603 | |||
AB445011.1 | - | 20 | 2152 | |||
AF115480.1 | - | 22 | 2142 | |||
AK004874.1 | - | 14 | 1260 | |||
AK220281.1 | - | 23 | 2268 | |||
BC139822.1 | - | 23 | 2200 | 40141811 |