Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019938.3 | NP_064322.1 | 11 | 1634 | ||
GenBank mRNA | AB029919.1 | - | 11 | 1634 | ||
AK006021.1 | - | 2 | 85 | |||
AK133053.1 | - | 10 | 1690 | |||
BC138245.1 | - | 11 | 1586 | 8861264 | ||
BC138246.1 | - | 11 | 1586 | 8861265 |