Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164107.1 | NP_001157579.1 | 7, 8 | 1115 | ||
NM_001164108.1 | NP_001157580.1 | 7, 8 | 1213 | |||
NM_019955.2 | NP_064339.2 | 7, 8 | 1132 | |||
GenBank mRNA | AA270784.1 | - | 2, 3 | 222 | ||
AF178953.1 | - | 7, 8 | 1061 | |||
AK012177.1 | - | 7, 8 | 1105 | |||
AK154505.1 | - | 7, 8 | 1125 | |||
BC029210.1 | - | 7, 8 | 1083 | 3590770 |