Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164195.1 | NP_001157667.1 | 1, 2 | 313 | ||
NM_019961.3 | NP_064345.1 | 1, 2 | 313 | |||
GenBank mRNA | AF152996.1 | - | 1, 2 | 236 | ||
AK012827.1 | - | 1, 2 | 292 | |||
AK075820.1 | - | 1, 2 | 228 | |||
AK146934.1 | - | 1, 2 | 292 | |||
BC033415.1 | - | 1, 2 | 195 | 5008068 | ||
BC037606.1 | - | 1, 2 | 276 | 5361347 |