Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177373.1 | NP_001170844.1 | 3 | 282 | ||
NM_019974.2 | NP_064358.1 | 2 | 248 | |||
GenBank mRNA | AB016226.1 | - | 3 | 132 | ||
AB016227.1 | - | 3 | 242 | |||
AK009360.1 | - | 3 | 283 | |||
AK009659.1 | - | 3 | 258 | |||
AK009720.1 | - | 3 | 283 | |||
BC115518.2 | - | 3 | 114 |