Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019827.6 | NP_062801.1 | 2 | 1691 | ||
GenBank mRNA | AB066114.1 | - | 2 | 1690 | ||
AF156099.2 | - | 2 | 332 | |||
AK154293.1 | - | 2 | 1169 | |||
AK164048.1 | - | 1 | 43 | |||
AK170965.1 | - | 2 | 1138 | |||
AK171482.1 | - | 2 | 1208 | |||
AK190946.1 | - | 2 | 135 | |||
BC006936.1 | - | 2 | 198 | 2648507 | ||
BC060743.1 | - | 2 | 1618 | 4022374 |