Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 4 | 830 | ||
GenBank mRNA | AF021833.1 | - | 1 | 185 | ||
AK049399.1 | - | 4 | 799 | |||
AK051044.1 | - | 2 | 465 | |||
AK083255.1 | - | 4 | 799 | |||
AK162966.1 | - | 4 | 831 | |||
BC054105.1 | - | 1 | 133 | 30022728 |