Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019977.2 | NP_064361.2 | 9, 10 | 770 | ||
GenBank mRNA | AF197127.1 | - | 9, 10 | 768 | ||
AK002406.1 | - | 9, 10 | 783 | |||
AY738257.1 | - | 8, 9 | 740 | |||
BC013543.1 | - | 9, 10 | 770 | 4242159 |