Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019981.2 | NP_064365.1 | 4 | 445 | ||
GenBank mRNA | AB022914.1 | - | 4 | 443 | ||
AK005769.1 | - | 4 | 439 | |||
BC048475.1 | - | 4 | 436 | 6740387 | ||
DQ222038.1 | - | 644 | ||||
DQ222039.1 | - | 582 | ||||
DQ222040.1 | - | 945 | ||||
DQ222041.1 | - | 886 | ||||
DQ222042.1 | - | 4 | 447 |