Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253916.1 | NP_001240845.1 | 2 | 560 | ||
NM_001253917.1 | NP_001240846.1 | 2 | 560 | |||
NM_019982.3 | NP_064366.2 | 2 | 560 | |||
GenBank mRNA | AB206790.1 | - | 2 | 539 | ||
AK083229.1 | - | 2 | 524 | |||
BC065117.1 | - | 2 | 520 | 6840338 | ||
BC065129.1 | - | 2 | 337 | 30362651 | ||
DQ167195.1 | - | 2 | 547 |