Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019992.3 | NP_064376.1 | 9 | 861 | ||
GenBank mRNA | AB036058.1 | - | 9 | 851 | ||
AK041474.1 | - | 9 | 862 | |||
BC145550.1 | - | 8 | 763 | 9054114 | ||
BC145828.1 | - | 9 | 876 | 40131131 | ||
BC145830.1 | - | 9 | 877 | 40131133 |