Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hacl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019975.3 | NP_064359.2 | 6 | 534 | ||
GenBank mRNA | AJ132139.1 | - | 6 | 532 | ||
AK005505.1 | - | 6 | 521 | |||
AK041686.1 | - | 6 | 530 | |||
AK050078.1 | - | 6 | 534 | |||
AK161488.1 | - | 514 | ||||
BC021360.1 | - | 6 | 465 | 5011451 |