Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174047.1 | NP_001167518.1 | 34, 35 | 3144 | ||
NM_001174048.1 | NP_001167519.1 | 33, 34 | 3126 | |||
NM_001174049.1 | NP_001167520.1 | 33, 34 | 3123 | |||
NM_001174050.1 | NP_001167521.1 | 33, 34 | 3126 | |||
NM_020263.3 | NP_064659.2 | 34, 35 | 3144 | |||
GenBank mRNA | AB093246.1 | - | 33, 34 | 2781 | ||
AF169633.1 | - | 2, 3 | 162 | |||
AF247139.1 | - | 35, 36 | 3398 | |||
AF247141.1 | - | 33, 34 | 2739 | |||
AK044603.1 | - | 33, 34 | 2942 | |||
AK161839.1 | - | 7, 8 | 597 | |||
AK164143.1 | - | 12, 13 | 970 | |||
AY502107.1 | - | 33, 34 | 3051 | |||
BC056389.1 | - | 34, 35 | 3144 | 6854935 | ||
BC158058.1 | - | 33, 34 | 2954 | 9007358 |