Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020277.2 | NP_064673.2 | 20 | 2971 | ||
GenBank mRNA | AB039952.1 | - | 19 | 2888 | ||
AF228681.1 | - | 20 | 2971 | |||
AJ271092.2 | - | 19 | 2884 | |||
AK035197.1 | - | 16 | 2501 | |||
AK044908.1 | - | 16 | 2586 | |||
AK161030.1 | - | 11 | 1564 | |||
AY280364.1 | - | 20 | 2935 | |||
AY280365.1 | - | 20 | 2935 | |||
BC133712.1 | - | 19 | 2888 | 40143371 |