Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289757.1 | NP_001276686.1 | 5 | 658 | ||
NM_021470.6 | NP_067445.3 | 5 | 630 | |||
GenBank mRNA | AF190664.1 | - | 2 | 392 | ||
AK005795.1 | - | 5 | 585 | |||
AK006379.1 | - | 5 | 574 | |||
AK016026.1 | - | 5 | 601 | |||
BC050964.1 | - | 5 | 573 | |||
BC061103.1 | - | 5 | 399 | |||
BC089493.1 | - | 5 | 573 | 6741898 |