Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039386.1 | NP_001034475.1 | 10 | 1296 | ||
NM_001039387.1 | NP_001034476.1 | 9 | 1290 | |||
NM_001177654.1 | NP_001171125.1 | 9 | 1290 | |||
NM_001177655.1 | NP_001171126.1 | 8 | 1200 | |||
NM_020276.3 | NP_064672.2 | 9 | 1227 | |||
GenBank mRNA | AF266508.2 | - | 8 | 1193 | ||
AJ278902.1 | - | 10 | 1074 | |||
AK004986.2 | - | 9 | 1287 | |||
AK019560.1 | - | 10 | 1263 | |||
AK053402.1 | - | 8 | 1147 | |||
AK158803.1 | - | 10 | 1026 | |||
AK165300.1 | - | 9 | 1235 | |||
BC006642.1 | - | 9 | 1248 | 3483536 |