Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039386.1 | NP_001034475.1 | 14 | 1601 | ||
NM_001039387.1 | NP_001034476.1 | 13 | 1595 | |||
NM_001177654.1 | NP_001171125.1 | 13 | 1595 | |||
NM_001177655.1 | NP_001171126.1 | 12 | 1505 | |||
NM_020276.3 | NP_064672.2 | 13 | 1532 | |||
GenBank mRNA | AF266508.2 | - | 12 | 1498 | ||
AJ278902.1 | - | 14 | 1379 | |||
AK004986.2 | - | 13 | 1592 | |||
AK019560.1 | - | 14 | 1568 | |||
AK053402.1 | - | 12 | 1452 | |||
AK158803.1 | - | 14 | 1331 | |||
AK165300.1 | - | 13 | 1540 | |||
BC006642.1 | - | 13 | 1553 | 3483536 |