Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019568.2 | NP_062514.2 | 3 | 618 | ||
GenBank mRNA | AB041614.1 | - | 3 | 618 | ||
AF144754.1 | - | 3 | 442 | |||
AF152377.1 | - | 3 | 368 | |||
AF192557.1 | - | 3 | 616 | |||
AF252873.1 | - | 3 | 616 | |||
AF352784.1 | - | 3 | 228 | |||
AF352785.1 | - | 3 | 228 | |||
AK004615.1 | - | 3 | 462 | |||
AK014351.1 | - | 3 | 408 | |||
AK076112.1 | - | 3 | 460 | |||
AK155356.1 | - | 3 | 456 | |||
AU035952.1 | - | 3 | 615 | |||
BC147274.1 | - | 3 | 362 | 9056321 | ||
BC147275.1 | - | 3 | 362 | 9056329 |