Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020603.2 | NP_065628.1 | 11 | 1274 | ||
GenBank mRNA | AK145068.1 | - | 11 | 1329 | ||
BC013527.1 | - | 11 | 1317 | |||
BC026431.1 | - | 8 | 898 | |||
BC046977.1 | - | 11 | 1274 | 6315172 | ||
KJ650211.1 | - | 11 | 1259 | |||
KJ663716.1 | - | 11 | 1127 |