Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020603.2 | NP_065628.1 | 8 | 789 | ||
GenBank mRNA | AK145068.1 | - | 8 | 844 | ||
BC013527.1 | - | 8 | 832 | |||
BC026431.1 | - | 5 | 413 | |||
BC046977.1 | - | 8 | 789 | 6315172 | ||
KJ650211.1 | - | 8 | 774 |