Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cramp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020608.3 | NP_065633.2 | 15, 16 | 3019 | ||
GenBank mRNA | AK089393.1 | - | 15, 16 | 3279 | ||
AK129356.1 | - | 6, 7 | 1195 | |||
AK147411.1 | - | 15, 16 | 3020 | |||
AK147449.1 | - | 15, 16 | 3020 |