Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020579.2 | NP_065604.2 | 4 | 763 | ||
GenBank mRNA | AF142671.1 | - | 4 | 703 | ||
AK155471.1 | - | 5 | 863 | |||
AK187407.1 | - | 1 | 65 | |||
BC004095.1 | - | 4 | 522 | |||
BC013619.1 | - | 4 | 736 | 3965561 | ||
BC021830.1 | - | 2 | 1559 |