Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020579.2 | NP_065604.2 | 8 | 1292 | ||
GenBank mRNA | AF142671.1 | - | 8 | 1232 | ||
AK155471.1 | - | 9 | 1392 | |||
BC004095.1 | - | 8 | 1049 | |||
BC013619.1 | - | 8 | 1265 | 3965561 | ||
BC021830.1 | - | 6 | 2088 | |||
BC035546.1 | - | 2 | 2583 | |||
CD363941.1 | - | 3 | 204 |
Gene Symbol: Ppox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134341.1 | - | 4149 |