Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020579.2 | NP_065604.2 | 3 | 430 | ||
GenBank mRNA | AF142671.1 | - | 3 | 370 | ||
AK155471.1 | - | 4 | 530 | |||
BC013619.1 | - | 3 | 403 | 3965561 | ||
BC021830.1 | - | 2 | 707 | |||
BU703242.1 | - | 3 | 429 |