Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020621.4 | NP_065646.1 | 3 | 1351 | ||
GenBank mRNA | AK076364.1 | - | 3 | 1352 | ||
BC119206.1 | - | 2 | 1133 | 8733955 | ||
BC120511.1 | - | 2 | 1133 | 40129434 |
Set Membership: |
G-protein coupled receptor |