Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020570.2 | NP_065595.2 | 3 | 262 | ||
GenBank mRNA | AK014963.1 | - | 2 | 178 | ||
AK020192.1 | - | 3 | 239 | |||
BC033417.1 | - | 3 | 262 | 5357630 | ||
Y17040.1 | - | 3 | 256 |