Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem183a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042485.1 | NP_001035950.1 | 1 | 195 | ||
NM_020588.2 | NP_065613.1 | 1 | 195 | |||
GenBank mRNA | AB041592.1 | - | 1 | 151 | ||
AK004922.2 | - | 1 | 176 | |||
AK006183.1 | - | 1 | 151 | |||
AK007779.1 | - | 1 | 174 | |||
AK152852.1 | - | 1 | 135 | |||
AK168331.1 | - | 1 | 137 | |||
BC003210.1 | - | 1 | 123 | 3156957 | ||
BY142931.1 | - | 1 | 203 | |||
CF897336.1 | - | 1 | 126 |