Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020578.3 | NP_065603.2 | 5 | 1405 | ||
GenBank mRNA | AF155883.1 | - | 5 | 1376 | ||
AF506002.1 | - | 5 | 1200 | |||
AK128964.1 | - | 5 | 1384 | |||
AK141265.1 | - | 5 | 1407 | |||
BC026392.1 | - | 2 | 286 | |||
BC046596.1 | - | 5 | 1149 | 6312263 |