Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021301.3 | NP_067276.2 | 14 | 1360 | ||
GenBank mRNA | AF111811.1 | - | 14 | 1360 | ||
AK128939.1 | - | 14 | 1193 | |||
BC138627.1 | - | 14 | 1317 | 8861647 | ||
BC138628.1 | - | 14 | 1317 | 8861648 |