Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021317.2 | NP_067292.2 | 1 | 396 | ||
GenBank mRNA | AB028855.1 | - | 1 | 396 | ||
AK006201.1 | - | 1 | 422 | |||
AK077186.1 | - | 1 | 425 | |||
BC048738.1 | - | 1 | 385 | |||
BC104396.1 | - | 1 | 343 | |||
BC104397.1 | - | 1 | 343 | |||
BC119478.2 | - | 1 | 343 |