Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021320.3 | NP_067295.2 | 4, 5 | 1388 | ||
GenBank mRNA | AF268066.1 | - | 4, 5 | 983 | ||
AF281278.1 | - | 4, 5 | 1236 | |||
BC119182.1 | - | 4, 5 | 1070 | 8733931 | ||
BC119184.1 | - | 4, 5 | 1070 | 8733933 | ||
CX205916.1 | - | 2, 3 | 199 |