Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301692.1 | NP_001288621.1 | 4 | 538 | ||
NM_001301694.1 | NP_001288623.1 | 4 | 488 | |||
NM_021340.4 | NP_067315.1 | 5 | 660 | |||
GenBank mRNA | AF076930.1 | - | 5 | 580 | ||
AK044450.1 | - | 5 | 612 | |||
AK044649.1 | - | 5 | 611 | |||
AK149272.1 | - | 5 | 611 | |||
BC082611.1 | - | 5 | 597 | |||
BC086689.1 | - | 5 | 587 | |||
BC119055.1 | - | 5 | 594 | 8733804 | ||
CK623457.1 | - | 4 | 419 | |||
CK626179.1 | - | 4 | 476 |
Set Membership: |
G-protein coupled receptor |