Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034029.1 | NP_001029201.1 | 10 | 955 | ||
NM_001034031.1 | NP_001029203.1 | 12 | 1030 | |||
NM_145826.5 | NP_665825.2 | 11 | 1382 | |||
GenBank mRNA | AF458068.1 | - | 11 | 1032 | ||
AK046302.1 | - | 11 | 1385 | |||
AK046713.1 | - | 11 | 1044 | |||
AK078876.1 | - | 3 | 1296 | |||
AK169498.1 | - | 11 | 1210 | |||
BC026737.1 | - | 1 | 126 | |||
BC069861.1 | - | 11 | 1059 | 6311568 | ||
DQ092339.1 | - | 11 | 971 | |||
DQ092340.1 | - | 10 | 955 | |||
DQ092341.1 | - | 12 | 1030 |